missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD133 Antibody (PROM/6316), Novus Biologicals™
Mouse Monoclonal Antibody
272.00€ - 508.00€
Specifications
| Antigen | CD133 |
|---|---|
| Clone | PROM/6316 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 μg/ml, Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml, SDS-Page |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18624726
|
Novus Biologicals
NBP3-07898 |
100 μg |
508.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639538
|
Novus Biologicals
NBP3-07898SS |
20 μg |
272.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
CD133 Monoclonal antibody specifically detects CD133 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), SDS-PageSpezifikation
| CD133 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Cellular Signaling, Core ESC Like Genes, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1 | |
| Recombinant chimericCD133 protein (around aa180-380 and aa612-765). This Mab recognizes both glycosylated and non-glycosylated extracellular domains of CD133. (Uniprot: O43490) | |
| Primary | |
| Store at 4°C. |
| PROM/6316 | |
| Western Blot 1-2 μg/ml, Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| O43490, O43490 | |
| 8842 | |
| IgG1 | |
| Protein A or G purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts