missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD133 Antibody (PROM/6316), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07898SS
This item is not returnable.
View return policy
Description
CD133 Monoclonal antibody specifically detects CD133 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page
Specifications
| CD133 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 μg/ml, Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml, SDS-Page | |
| O43490, O43490 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PROM/6316 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1 | |
| Recombinant chimericCD133 protein (around aa180-380 and aa612-765). This Mab recognizes both glycosylated and non-glycosylated extracellular domains of CD133. (Uniprot: O43490) | |
| 20 μg | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Cellular Signaling, Core ESC Like Genes, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
| 8842 | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction