Learn More
CD117/c-kit Antibody (ACK2) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 5 publications
Brand:  Novus Biologicals NBP1-43359-0.1mg
 View more versions of this product
Product Code. 18758663
  • ONLINE EXCLUSIVE PRICE
    473.00€
    447.30€ / 0.10mg
    Save 25.70€
    5% Off
Description
CD117/c-kit Monoclonal specifically detects CD117/c-kit in Human, Mouse, Guinea Pig samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD117/c-kit
Monoclonal
0.5 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry < 0.125 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional, Immunohistochemistry Whole-Mount
P10721
KIT
Mouse CD117
0.1 mg
Cancer, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases
3815
Human, Mouse, Guinea Pig
IgG2b κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay
ACK2
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein
Rat
Affinity Purified
RUO
Primary
The ACK2 monoclonal antibody reacts with mouse CD117, also known as c-Kit receptor, Steel factor receptor and stem cell factor receptor.
Store at 4C. Do not freeze.