missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117/c-kit Antibody (ACK2) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 5 publications
215.00€ - 473.00€
Specifications
| Antigen | CD117/c-kit |
|---|---|
| Clone | ACK2 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry < 0.125 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional, Immunohistochemistry Whole-Mount |
| Applications | Immunohistochemistry, Functional Assay, Immunoprecipitation, Flow Cytometry, Western Blot, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18285064
|
Novus Biologicals
NBP1-43359-0.025MG |
0.025 mg |
215.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18777023
|
Novus Biologicals
NBP1-43359-0.05mg |
0.05 mg |
349.00€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18758663
|
Novus Biologicals
NBP1-43359-0.1mg |
0.1 mg |
473.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD117/c-kit Monoclonal specifically detects CD117/c-kit in Human, Mouse, Guinea Pig samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount.Specifications
| CD117/c-kit | |
| 0.5 mg/mL | |
| Immunohistochemistry, Functional Assay, Immunoprecipitation, Flow Cytometry, Western Blot, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Mouse, Human, Guinea Pig | |
| P10721 | |
| 3815 | |
| Mouse CD117 | |
| Primary | |
| Store at 4C. Do not freeze. |
| ACK2 | |
| Western Blot 1:100-1:2000, Flow Cytometry < 0.125 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional, Immunohistochemistry Whole-Mount | |
| Monoclonal | |
| Rat | |
| Cancer, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
| KIT | |
| IgG2b κ | |
| Affinity Purified | |
| The ACK2 monoclonal antibody reacts with mouse CD117, also known as c-Kit receptor, Steel factor receptor and stem cell factor receptor. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title