missing translation for 'onlineSavingsMsg'
Learn More

CCR11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

483.00€

Specifications

Antigen CCR11
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18257575
View Documents
Novus Biologicals
NBP1-79925
100 μL
483.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CCR11 Polyclonal specifically detects CCR11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CCR11
Polyclonal
Rabbit
NP_057641
51554
IgG
39 kDa
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
CC chemokine receptor-like 1, C-C CKR-11, CCBP2CCX CKR, CCR11CCR10, chemokine (C-C motif) receptor-like 1, orphan seven-transmembrane receptor, chemokine related, PPR1cc motif, receptor-like protein 1, VSHK1CKR-11
The specific Immunogen is proprietary information. Peptide sequence SIALFHSCLNPILYVFMGASFKNYVMKVAKKYGSWRRQRQSVEEFPFDSE.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.