missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL11/Eotaxin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00€
Specifications
| Antigen | CCL11/Eotaxin |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5-10 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
CCL11/Eotaxin Polyclonal specifically detects CCL11/Eotaxin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| CCL11/Eotaxin | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5-10 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Asthma, Cancer, Chemokine Research, Chemokines and Cytokines, Cytokine Research, Immunology | |
| PBS with 0.02% Sodium Azide | |
| C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin) | |
| CCL11 | |
| IgG | |
| Ion exchange chromatography | |
| 9-18 kDa |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| CAB07027 | |
| 6356 | |
| Antibody was raised in rabbits against a peptide corresponding to amino acids near the carboxy terminus of human eotaxin. The immunogen is located within the last 50 amino acids of Eotaxin. Amino Acid Squence: DSMKYLDQKSPTPKP | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title