missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL11/Eotaxin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77046-0.025mg
This item is not returnable.
View return policy
Description
CCL11/Eotaxin Polyclonal specifically detects CCL11/Eotaxin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CCL11/Eotaxin | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin) | |
| Rabbit | |
| 9-18 kDa | |
| 0.025 mg | |
| Asthma, Cancer, Chemokine Research, Chemokines and Cytokines, Cytokine Research, Immunology | |
| 6356 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5-10 μg/mL | |
| CAB07027 | |
| CCL11 | |
| Antibody was raised in rabbits against a peptide corresponding to amino acids near the carboxy terminus of human eotaxin. The immunogen is located within the last 50 amino acids of Eotaxin. Amino Acid Squence: DSMKYLDQKSPTPKP | |
| Ion exchange chromatography | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction