missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CBF2 Antibody (PCRP-CEBPZ-2D8), Novus Biologicals™
Mouse Monoclonal Antibody
232.00€ - 462.00€
Specifications
| Antigen | CBF2 |
|---|---|
| Clone | PCRP-CEBPZ-2D8 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1-2 μg/million cells, Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043051
|
Novus Biologicals™
NBP3-26762-20ug |
20 μg |
232.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043369
|
Novus Biologicals™
NBP3-26762-100ug |
100 μg |
462.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CBF2 Monoclonal antibody specifically detects CBF2 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin)Specifications
| CBF2 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| CBF2CTF2, CBFHSP-CBF, CCAAT/enhancer binding protein (C/EBP), zeta, CCAAT/enhancer binding protein zeta, CCAAT/enhancer-binding protein zeta, CCAAT-binding factor, CCAAT-box-binding transcription factor, NOC1 | |
| Protein domain of CBF2 protein | |
| Primary | |
| Store at 4°C. |
| PCRP-CEBPZ-2D8 | |
| Flow Cytometry 1-2 μg/million cells, Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 10153 | |
| IgG2c | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title