missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CBF2 Antibody (PCRP-CEBPZ-2D8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26762-100ug
This item is not returnable.
View return policy
Description
CBF2 Monoclonal antibody specifically detects CBF2 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin)
Specifications
| CBF2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1-2 μg/million cells, Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| CBF2CTF2, CBFHSP-CBF, CCAAT/enhancer binding protein (C/EBP), zeta, CCAAT/enhancer binding protein zeta, CCAAT/enhancer-binding protein zeta, CCAAT-binding factor, CCAAT-box-binding transcription factor, NOC1 | |
| Protein domain of CBF2 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| PCRP-CEBPZ-2D8 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 10153 | |
| Store at 4°C. | |
| IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction