missing translation for 'onlineSavingsMsg'
Learn More

Cav1.3 Antibody (S48), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

174.00€

Specifications

Antigen Cav1.3
Clone S48
Concentration 1 mg/mL
Dilution Western Blot 1 - 10 μg/mL, Immunohistochemistry 0.1 - 1 μg/mL, Immunocytochemistry/Immunofluorescence 1 - 10 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Microarray
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18782994
View Documents
Novus Biologicals
NBP2-12893-0.025mg
0.025 mg
174.00€
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cav1.3 Monoclonal specifically detects Cav1.3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.
Specifications

Specifications

Cav1.3
1 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay
Unconjugated
Mouse
Human, Mouse, Rat
776
Fusion protein amino acids 859-875 of rat Cav1.3
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
S48
Western Blot 1 - 10 μg/mL, Immunohistochemistry 0.1 - 1 μg/mL, Immunocytochemistry/Immunofluorescence 1 - 10 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Microarray
Monoclonal
Purified
RUO
CACN4calcium channel, neuroendocrine/brain-type, alpha 1 subunit, CACNL1A2voltage-dependent L-type calcium channel subunit alpha-1D, Calcium channel, L type, alpha-1 polypeptide, isoform 2, calcium channel, voltage-dependent, L type, alpha 1D subunit, Cav1.3, CCHL1A2voltage-gated calcium channel alpha 1 subunit, L type, alpha-1 polypeptide, voltage-gated calcium channel alpha subunit Cav1.3, Voltage-gated calcium channel subunit alpha Cav1.3
CACNA1D
IgG2a κ
Protein G purified
Detects approx 250 kDa. No cross-reactivity against Cav1.2.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.