Learn More
Cav1.3 Antibody (S48), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-12893-0.025mg
This item is not returnable.
View return policy
Description
Cav1.3 Monoclonal specifically detects Cav1.3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.
Specifications
| Cav1.3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 - 10 μg/mL, Immunohistochemistry 0.1 - 1 μg/mL, Immunocytochemistry/Immunofluorescence 1 - 10 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Microarray | |
| CACNA1D | |
| Fusion protein amino acids 859-875 of rat Cav1.3 | |
| 0.025 mg | |
| Primary | |
| Detects approx 250 kDa. No cross-reactivity against Cav1.2. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay | |
| S48 | |
| Unconjugated | |
| CACN4calcium channel, neuroendocrine/brain-type, alpha 1 subunit, CACNL1A2voltage-dependent L-type calcium channel subunit alpha-1D, Calcium channel, L type, alpha-1 polypeptide, isoform 2, calcium channel, voltage-dependent, L type, alpha 1D subunit, Cav1.3, CCHL1A2voltage-gated calcium channel alpha 1 subunit, L type, alpha-1 polypeptide, voltage-gated calcium channel alpha subunit Cav1.3, Voltage-gated calcium channel subunit alpha Cav1.3 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 776 | |
| Human, Mouse, Rat | |
| Purified |