missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin X/Z/P Antibody (6G11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
339.00€ - 468.00€
Specifications
| Antigen | Cathepsin Z |
|---|---|
| Clone | 6G11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA 1:1000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18113323
|
Novus Biologicals
NBP2-31211-0.05ML |
0.05 mL |
339.00€
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18188102
|
Novus Biologicals
NBP2-31211-0.1ML |
0.1 mL |
468.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cathepsin X/Z/P Monoclonal specifically detects Cathepsin X/Z/P in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Cathepsin Z | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_001327 | |
| 1522 | |
| Recombinant human CTSZ (62-303aa) purified from E. coli | |
| Primary | |
| Protein A purified |
| 6G11 | |
| Western Blot 1:1000, ELISA 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide | |
| Cathepsin P, Cathepsin X, cathepsin Z, CTSX, EC 3.4.18.1, FLJ17088, preprocathepsin P | |
| CTSZ | |
| IgG2b κ | |
| Protein A purified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title