missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin X/Z/P Antibody (6G11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-31211-0.1ML
This item is not returnable.
View return policy
Description
Cathepsin X/Z/P Monoclonal specifically detects Cathepsin X/Z/P in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Cathepsin Z | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA 1:1000 | |
| NP_001327 | |
| CTSZ | |
| Recombinant human CTSZ (62-303aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Protein A purified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA | |
| 6G11 | |
| Unconjugated | |
| PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide | |
| Cathepsin P, Cathepsin X, cathepsin Z, CTSX, EC 3.4.18.1, FLJ17088, preprocathepsin P | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 1522 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction