missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin K Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
202.00€ - 573.00€
Specifications
| Antigen | Cathepsin K |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30226545
|
Novus Biologicals
NBP3-35793-100ul |
100 μL |
573.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228581
|
Novus Biologicals
NBP3-35793-20ul |
20 μL |
202.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cathepsin K Polyclonal antibody specifically detects Cathepsin K in Human,Mouse samples. It is validated for ELISA,Western BlotSpecifications
| Cathepsin K | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.3), 50% glycerol | |
| 1513 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| cathepsin K, cathepsin K (pycnodysostosis), Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin X, CTS02, CTSO, CTSO1, CTSO2, EC 3.4.22, EC 3.4.22.38, MGC23107, PKND, PYCD | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 115-329 of human Cathepsin K (NP_000387.1).,, Sequence:, APDSVDYRKKGYVTPVKNQGQCGSCWAFSSVGALEGQLKKKTGKLLNLSPQNLVDCVSENDGCGGGYMTNAFQYVQKNRGIDSEDAYPYVGQEESCMYNPTGKAAKCRGYREIPEGNEKALKRAVARVGPVSVAIDASLTSFQFYSKGVYYDESCNSDNLNHAVLAVGYGIQKGNKHWIIKNSWGENWGNKGYILMARNKNNACGIANLASFPKM | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title