Learn More
Cathepsin K Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35793-100ul
This item is not returnable.
View return policy
Description
Cathepsin K Polyclonal antibody specifically detects Cathepsin K in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Cathepsin K | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, ELISA | |
| cathepsin K, cathepsin K (pycnodysostosis), Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin X, CTS02, CTSO, CTSO1, CTSO2, EC 3.4.22, EC 3.4.22.38, MGC23107, PKND, PYCD | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 115-329 of human Cathepsin K (NP_000387.1).,, Sequence:, APDSVDYRKKGYVTPVKNQGQCGSCWAFSSVGALEGQLKKKTGKLLNLSPQNLVDCVSENDGCGGGYMTNAFQYVQKNRGIDSEDAYPYVGQEESCMYNPTGKAAKCRGYREIPEGNEKALKRAVARVGPVSVAIDASLTSFQFYSKGVYYDESCNSDNLNHAVLAVGYGIQKGNKHWIIKNSWGENWGNKGYILMARNKNNACGIANLASFPKM | |
| 100 μL | |
| Cancer | |
| 1513 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |