missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARD14 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
222.00€ - 531.00€
Specifications
| Antigen | CARD14 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615162
|
Novus Biologicals
NBP2-92873-0.02ml |
0.02 mL |
222.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661792
|
Novus Biologicals
NBP2-92873-0.1ml |
0.1 mL |
531.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CARD14 Polyclonal antibody specifically detects CARD14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CARD14 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS with 50% glycerol, pH7.3. | |
| 79092 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| bcl10-interacting maguk protein 2, BIMP2, CARD-containing MAGUK protein 2, card-maguk protein 2, Carma 2, CARMA2CARD-containing MAGUK 2 protein, caspase recruitment domain family, member 14, caspase recruitment domain-containing protein 14 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-500 of human CARD14 (NP_001244899.1). SLSLHYSNALQEKELAASRCRSLQEELYLLKQELQRANMVSSCELELQEQSLRTASDQESGDEELNRLKEENEKLRSLTFSLAEKDILEQSLDEARGSRQELVERIHSLRERAVAAERQREQYWEEKEQTLLQFQKSKMACQLYREKVNALQAQVCELQKERDQAYSARDSAQREISQSLVEKDSLRRQVFELTDQVCELRTQLRQLQAEPPGVLKQEARTREPCPREKQRLVRMHAICPRDDSDCSLVSSTESQLLSDLSATSSRELVDSFRSSSPAPPSQQSLYKRVAEDFGEEPWSFS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title