missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARD14 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92873-0.02ml
This item is not returnable.
View return policy
Description
CARD14 Polyclonal antibody specifically detects CARD14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CARD14 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| bcl10-interacting maguk protein 2, BIMP2, CARD-containing MAGUK protein 2, card-maguk protein 2, Carma 2, CARMA2CARD-containing MAGUK 2 protein, caspase recruitment domain family, member 14, caspase recruitment domain-containing protein 14 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-500 of human CARD14 (NP_001244899.1). SLSLHYSNALQEKELAASRCRSLQEELYLLKQELQRANMVSSCELELQEQSLRTASDQESGDEELNRLKEENEKLRSLTFSLAEKDILEQSLDEARGSRQELVERIHSLRERAVAAERQREQYWEEKEQTLLQFQKSKMACQLYREKVNALQAQVCELQKERDQAYSARDSAQREISQSLVEKDSLRRQVFELTDQVCELRTQLRQLQAEPPGVLKQEARTREPCPREKQRLVRMHAICPRDDSDCSLVSSTESQLLSDLSATSSRELVDSFRSSSPAPPSQQSLYKRVAEDFGEEPWSFS | |
| 0.02 mL | |
| Apoptosis | |
| 79092 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction