missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcineurin A Mouse anti-Human, Clone: BF1.1, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | Calcineurin A |
|---|---|
| Clone | BF1.1 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, ELISA |
| Applications | Western Blot, ELISA |
Description
Calcineurin A Monoclonal specifically detects Calcineurin A in Human samples. It is validated for Western Blot, ELISA.Specifications
| Calcineurin A | |
| 0.2 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Immunology, Protein Phosphatase, Wnt Signaling Pathway | |
| 10 mM PBS with 0.05% BSA | |
| 5530 | |
| IgG1 κ | |
| Protein A or G purified |
| BF1.1 | |
| Western Blot 1 to 2 μg/mL, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| calcineurin A alpha, Calmodulin-dependent calcineurin A subunit alpha isoform, CALN, CALNAcatalytic subunit, alpha isoform, CAM-PRP catalytic subunit, CNA, CNA1CALNA1, EC 3.1.3.16, PPP2BCCN1, protein phosphatase 2B, catalytic subunit, alpha isoform, protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform(calcineurin A alpha), protein phosphatase 3, catalytic subunit, alpha isozyme, serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform | |
| Recombinant full-length Calcineurin A protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title