missing translation for 'onlineSavingsMsg'
Learn More

Calcineurin A Mouse anti-Human, Clone: BF1.1, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-07657-100ug

 View more versions of this product

Product Code. 18629857

  • 484.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Calcineurin A Monoclonal specifically detects Calcineurin A in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calcineurin A
Monoclonal
0.2 mg/mL
Western Blot 1 to 2 μg/mL, ELISA
calcineurin A alpha, Calmodulin-dependent calcineurin A subunit alpha isoform, CALN, CALNAcatalytic subunit, alpha isoform, CAM-PRP catalytic subunit, CNA, CNA1CALNA1, EC 3.1.3.16, PPP2BCCN1, protein phosphatase 2B, catalytic subunit, alpha isoform, protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform(calcineurin A alpha), protein phosphatase 3, catalytic subunit, alpha isozyme, serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform
Recombinant full-length Calcineurin A protein
100 μg
Adaptive Immunity, Immunology, Protein Phosphatase, Wnt Signaling Pathway
5530
Store at 4°C.
IgG1 κ
Western Blot, ELISA
BF1.1
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.