missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CACNB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30830-25ug
This item is not returnable.
View return policy
Description
CACNB1 Polyclonal antibody specifically detects CACNB1 in Human samples. It is validated for Western Blot
Specifications
| CACNB1 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL | |
| CAB1, CACNLB1calcium channel, L type, beta 1 polypeptide, Calcium channel voltage-dependent subunit beta 1, calcium channel, voltage-dependent, beta 1 subunit, CCHLB1, dihydropyridine-sensitive L-type, calcium channel beta-1 subunit, MGC41896, voltage-dependent L-type calcium channel subunit beta-1 | |
| Synthetic peptide derived from the rat CACNB1 conjugated to KLH | |
| 25 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 782 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction