missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CACNB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00€ - 508.00€
Specifications
| Antigen | CACNB1 |
|---|---|
| Dilution | Western Blot : 5-10 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067150
|
Novus Biologicals™
NBP3-30830-100ug |
100 μg |
508.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066980
|
Novus Biologicals™
NBP3-30830-25ug |
25 μg |
213.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CACNB1 Polyclonal antibody specifically detects CACNB1 in Human samples. It is validated for Western BlotSpecifications
| CACNB1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CAB1, CACNLB1calcium channel, L type, beta 1 polypeptide, Calcium channel voltage-dependent subunit beta 1, calcium channel, voltage-dependent, beta 1 subunit, CCHLB1, dihydropyridine-sensitive L-type, calcium channel beta-1 subunit, MGC41896, voltage-dependent L-type calcium channel subunit beta-1 | |
| Synthetic peptide derived from the rat CACNB1 conjugated to KLH | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 5-10 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline | |
| 782 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title