Learn More
Bupivacaine, Thermo Scientific™
Bupivacaine, CAS # 38396-39-3, is a potent and long-acting in vivo local anesthetic agent.
102.00€ - 803.00€
Chemical Identifiers
| CAS | 38396-39-3 |
|---|---|
| Molecular Formula | C18H28N2O |
| Molecular Weight (g/mol) | 288.44 |
| MDL Number | MFCD00243007,MFCD00941462 |
| InChI Key | LEBVLXFERQHONN-UHFFFAOYNA-N |
| Synonym | bupivacaine, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide, dl-bupivacaine, marcaine, bupivacaina, carbostesin, bupivacainum, sensorcaine, anekain, +--bupivacaine |
| PubChem CID | 2474 |
| ChEBI | CHEBI:77431 |
| IUPAC Name | 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide |
| SMILES | CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15485049
|
Thermo Scientific Alfa Aesar
J62742.03 |
1g |
102.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15495049
|
Thermo Scientific Alfa Aesar
J62742.06 |
5g |
176.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15405059
|
Thermo Scientific Alfa Aesar
J62742.14 |
25g |
803.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 38396-39-3 | |
| 288.44 | |
| LEBVLXFERQHONN-UHFFFAOYNA-N | |
| 2474 | |
| 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide |
| C18H28N2O | |
| MFCD00243007,MFCD00941462 | |
| bupivacaine, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide, dl-bupivacaine, marcaine, bupivacaina, carbostesin, bupivacainum, sensorcaine, anekain, +--bupivacaine | |
| CHEBI:77431 | |
| CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C |
Specifications
| 38396-39-3 | |
| MFCD00243007,MFCD00941462 | |
| bupivacaine, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide, dl-bupivacaine, marcaine, bupivacaina, carbostesin, bupivacainum, sensorcaine, anekain, +--bupivacaine | |
| LEBVLXFERQHONN-UHFFFAOYNA-N | |
| 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide | |
| 2474 | |
| 288.43 |
| C18H28N2O | |
| UN2811 | |
| Soluble in water,ethanol,chloroform (slightly ),and acetone (slightly ). | |
| CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C | |
| 288.44 | |
| CHEBI:77431 | |
| Bupivacaine |
Safety and Handling
GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
H300+H310+H330
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 253-911-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.