Learn More
Bupivacaine, Thermo Scientific™
Bupivacaine, CAS # 38396-39-3, is a potent and long-acting in vivo local anesthetic agent.
Brand: Thermo Scientific Alfa Aesar J62742.06
| Quantity | 5g |
|---|
View more versions of this product
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 38396-39-3 | |
| 288.44 | |
| LEBVLXFERQHONN-UHFFFAOYNA-N | |
| 2474 | |
| 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide |
| C18H28N2O | |
| MFCD00243007,MFCD00941462 | |
| bupivacaine, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide, dl-bupivacaine, marcaine, bupivacaina, carbostesin, bupivacainum, sensorcaine, anekain, +--bupivacaine | |
| CHEBI:77431 | |
| CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C |
Specifications
| 38396-39-3 | |
| MFCD00243007,MFCD00941462 | |
| UN2811 | |
| Soluble in water,ethanol,chloroform (slightly ),and acetone (slightly ). | |
| CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C | |
| 288.44 | |
| CHEBI:77431 | |
| Bupivacaine |
| C18H28N2O | |
| 5g | |
| bupivacaine, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide, dl-bupivacaine, marcaine, bupivacaina, carbostesin, bupivacainum, sensorcaine, anekain, +--bupivacaine | |
| LEBVLXFERQHONN-UHFFFAOYNA-N | |
| 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide | |
| 2474 | |
| 288.43 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.