missing translation for 'onlineSavingsMsg'
Learn More

Bupivacaine, Thermo Scientific™

Bupivacaine, CAS # 38396-39-3, is a potent and long-acting in vivo local anesthetic agent.

Brand:  Thermo Scientific Alfa Aesar J62742.03

Quantity 1g

 View more versions of this product

Product Code. 15485049

  • 102.00€ / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Bupivacaine is a local anesthetic agent
• The compound acts by blocking sodium influx into neurons through intracellular binding at voltage-gated sodium channels
• Bupivacaine also inhibits NMDA receptors on nerve cells, resulting in reduced synaptic transmission
Application
• Bupivacaine can be used in the laboratory to study treatment of acute and chronic pain in a preclinical setting
• Bupivacaine is used as a local anesthetic in rodent surgery in laboratory settings
• Bupivacaine has been shown to activate the AMPKα/BDNF signaling pathway in spinal astrocytes
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

38396-39-3
288.44
LEBVLXFERQHONN-UHFFFAOYNA-N
2474
1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide
C18H28N2O
MFCD00243007,MFCD00941462
bupivacaine, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide, dl-bupivacaine, marcaine, bupivacaina, carbostesin, bupivacainum, sensorcaine, anekain, +--bupivacaine
CHEBI:77431
CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C

Specifications

38396-39-3
MFCD00243007,MFCD00941462
UN2811
Soluble in water,ethanol,chloroform (slightly ),and acetone (slightly ).
CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C
288.44
CHEBI:77431
Bupivacaine
C18H28N2O
1g
bupivacaine, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide, dl-bupivacaine, marcaine, bupivacaina, carbostesin, bupivacainum, sensorcaine, anekain, +--bupivacaine
LEBVLXFERQHONN-UHFFFAOYNA-N
1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide
2474
288.43
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.