missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ BubR1 Antibody - N-terminal, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30806-250ug
This item is not returnable.
View return policy
Description
BubR1 Polyclonal antibody specifically detects BubR1 in Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| BubR1 | |
| Polyclonal | |
| Western Blot : 1-10 μg/mL, Immunohistochemistry, Immunoprecipitation : 1-10 μg/mL | |
| Bub1A, BUBR1budding uninhibited by benzimidazoles 1 (yeast homolog), beta, budding uninhibited by benzimidazoles 1 homolog beta (yeast), EC 2.7.11.1, hBUBR1, MAD3/BUB1-related protein kinase, MAD3LBUB1beta, Mitotic checkpoint kinase MAD3L, Protein SSK1, SSK1mitotic checkpoint serine/threonine-protein kinase BUB1 beta | |
| Recombinant protein corresponding to amino acids 10-190 of the mouse BubR1 protein. | |
| 250 μg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
| 701 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Sheep | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction