missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ BubR1 Antibody - N-terminal, Novus Biologicals™

Sheep Polyclonal Antibody

213.00€ - 508.00€

Specifications

Antigen BubR1
Dilution Western Blot : 1-10 μg/mL, Immunohistochemistry, Immunoprecipitation : 1-10 μg/mL
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30066791
View Documents
Novus Biologicals™
NBP3-30806-50ug
50 μg
213.00€
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30066958
View Documents
Novus Biologicals™
NBP3-30806-250ug
250 μg
508.00€
250µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BubR1 Polyclonal antibody specifically detects BubR1 in Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications

Specifications

BubR1
Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Sheep
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers
Phosphate buffered saline
701
IgG
Ammonium sulfate precipitation
Western Blot : 1-10 μg/mL, Immunohistochemistry, Immunoprecipitation : 1-10 μg/mL
Polyclonal
Purified
RUO
Mouse
Bub1A, BUBR1budding uninhibited by benzimidazoles 1 (yeast homolog), beta, budding uninhibited by benzimidazoles 1 homolog beta (yeast), EC 2.7.11.1, hBUBR1, MAD3/BUB1-related protein kinase, MAD3LBUB1beta, Mitotic checkpoint kinase MAD3L, Protein SSK1, SSK1mitotic checkpoint serine/threonine-protein kinase BUB1 beta
Recombinant protein corresponding to amino acids 10-190 of the mouse BubR1 protein.
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.