Learn More
Macherey-Nagel™ BTX Test Mixture in Methanol
Product Code. 10033001 Shop All Macherey-Nagel Products
Change view
Click to view available options
Type:
BTX Test Mixture in Methanol
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10033001 BTX Test Mixture in Methanol 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10033001 Supplier Macherey-Nagel™ Supplier No. 722372
 Restricted Item
Estimated Shipment: 24-12-2026
Specifications
Analyte or Component Names Benzene, Ethylbenzene, Toluene, m-, o-, p-xylene
Technique GC Environmental Analyses
Type BTX Test Mixture in Methanol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • BTX test mixture, conc. 10 ng/
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: BTX test mixture inmethanol