missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTAF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-57414
This item is not returnable.
View return policy
Description
BTAF1 Polyclonal specifically detects BTAF1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| BTAF1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| ATP-dependent helicase BTAF1, BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa (Mot1homolog, S. cerevisiae), B-TFIID transcription factor-associated 170 kDa subunit, EC 3.6.1, EC 3.6.4.-, MOT1MGC138406, TAF(II)170KIAA0940, TAF-172, TAF172BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170 kD (Mot1homolog, S. cerevisiae), TAFII170TATA-binding protein-associated factor 172, TBP-associated factor 172 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9044 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| BTAF1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QFAARYGKPILASRDARSSSREQEAGVLAMDALHRQVLPFLLRRMKEDVLQDLPPKIIQDYYCTLSPLQVQLYEDFAKSRAKCDVDETVSSATLS | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction