missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bovine Serum Albumin Antibody (D1) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
244.00€ - 635.00€
Specifications
| Antigen | Bovine Serum Albumin |
|---|---|
| Clone | D1 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18628454
|
Novus Biologicals
NBP3-20114-0.025mg |
0.025 mg |
244.00€
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18630904
|
Novus Biologicals
NBP3-20114-0.2mg |
0.2 mg |
635.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bovine Serum Albumin Monoclonal antibody specifically detects Bovine Serum Albumin in Bovine samples. It is validated for ELISA, ImmunoprecipitationSpecifications
| Bovine Serum Albumin | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Bovine | |
| albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| D1 was prepared by isolating anti-p24 scFvs from a phage display library made from a mouse immunized with lysate from HIV-1 infected H9 cells. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| D1 | |
| ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers | |
| PBS | |
| 280717 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title