missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bovine Serum Albumin Antibody (D1) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20114-0.2mg
This item is not returnable.
View return policy
Description
Bovine Serum Albumin Monoclonal antibody specifically detects Bovine Serum Albumin in Bovine samples. It is validated for ELISA, Immunoprecipitation
Specifications
| Bovine Serum Albumin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Bovine | |
| Purified |
| ELISA, Immunoprecipitation | |
| D1 | |
| Unconjugated | |
| albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| D1 was prepared by isolating anti-p24 scFvs from a phage display library made from a mouse immunized with lysate from HIV-1 infected H9 cells. | |
| 0.2 mg | |
| Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers | |
| 280717 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction