missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLZF1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | BLZF1 |
|---|---|
| Concentration | 350 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236274
|
Proteintech
16354-1-AP-20UL |
20 μL |
145.00€ 129.00€ / 20µL Save 16.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233906
|
Proteintech
16354-1-AP-150UL |
150 μL |
410.00€ 349.00€ / 150µL Save 61.00€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The BLZF1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human BLZF1. This antibody recognizes human, mouse antigen. The BLZF1 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| BLZF1 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| GOLGIN-45; JEM-1; JEM-1s; JEM1; MGC22497 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 350 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 8548 | |
| Fusion Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts