Learn More
BLZF1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16354-1-AP-150UL
This item is not returnable.
View return policy
Description
The BLZF1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human BLZF1. This antibody recognizes human, mouse antigen. The BLZF1 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| BLZF1 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8548 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 350 μg/mL | |
| GOLGIN-45; JEM-1; JEM-1s; JEM1; MGC22497 | |
| Fusion Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| 45 kDa | |
| Liquid |