missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin-conjugated CD9 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech BIOTIN-60232-100UL
This item is not returnable.
View return policy
Description
The CD9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD9. This antibody recognizes human antigen. The CD9 antibody has been validated for the following applications: IHC analysis. This antibody is biotin.Specifications
| CD9 | |
| Monoclonal | |
| 1000 μg/mL | |
| 5H9; BA2; BTCC-1; DRAP-27; GIG2; MIC3; MRP-1; P24; TSPAN29 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human | |
| 23-27 kDa | |
| Liquid |
| Immunohistochemistry | |
| 4H7B9 | |
| Biotin | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 928 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction