missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin-conjugated CD9 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | CD9 |
|---|---|
| Clone | 4H7B9 |
| Concentration | 1000 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237109
|
Proteintech
BIOTIN-60232-20UL |
20 μL |
140.00€ 129.00€ / 20µL Save 11.00€ 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237192
|
Proteintech
BIOTIN-60232-100UL |
100 μL |
450.00€ 349.00€ / 100µL Save 101.00€ 22% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD9. This antibody recognizes human antigen. The CD9 antibody has been validated for the following applications: IHC analysis. This antibody is biotin.Specifications
| CD9 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 928 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 4H7B9 | |
| Immunohistochemistry | |
| Biotin | |
| Mouse | |
| Human | |
| 5H9; BA2; BTCC-1; DRAP-27; GIG2; MIC3; MRP-1; P24; TSPAN29 | |
| IgG1 | |
| Protein G purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title