missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biliverdin Reductase A/BLVRA Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
195.00€
Specifications
| Antigen | Biliverdin Reductase A/BLVRA |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.1 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18547171
|
Novus Biologicals
NB120-3466-0.025mg |
0.025 mg | |||||||
Description
Biliverdin Reductase A/BLVRA Polyclonal specifically detects Biliverdin Reductase A/BLVRA in Human samples. It is validated for Western Blot.Specifications
| Biliverdin Reductase A/BLVRA | |
| Western Blot 0.1 μg/mL | |
| Polyclonal | |
| Chicken | |
| Human | |
| biliverdin reductase A, Biliverdin-IX alpha-reductase, BLVR, BVR, BVR A, BVRA, EC 1.3.1.24 | |
| BLVRA | |
| IgY | |
| Affinity Purified | |
| NB120-3466 detects biliverdin-IX alpha reductase (BLVRA) from human cells. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 644 | |
| Purified human biliverdin-IX alpha-reductase expressed in E. coli. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title