missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biliverdin Reductase A/BLVRA Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB120-3466-0.025mg
This item is not returnable.
View return policy
Description
Biliverdin Reductase A/BLVRA Polyclonal specifically detects Biliverdin Reductase A/BLVRA in Human samples. It is validated for Western Blot.
Specifications
| Biliverdin Reductase A/BLVRA | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| BLVRA | |
| Purified human biliverdin-IX alpha-reductase expressed in E. coli. | |
| 0.025 mg | |
| Primary | |
| NB120-3466 detects biliverdin-IX alpha reductase (BLVRA) from human cells. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.1 μg/mL | |
| biliverdin reductase A, Biliverdin-IX alpha-reductase, BLVR, BVR, BVR A, BVRA, EC 1.3.1.24 | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| 644 | |
| Human | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction