missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Tubulin Antibody (TUBB/4470), Novus Biologicals™
Mouse Monoclonal Antibody
249.00€ - 494.00€
Specifications
| Antigen | beta Tubulin |
|---|---|
| Clone | TUBB/4470 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18698814
|
Novus Biologicals
NBP3-20588-20ug |
20 μg |
249.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640394
|
Novus Biologicals
NBP3-20588-100ug |
100 μg |
494.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta Tubulin Monoclonal antibody specifically detects beta Tubulin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| beta Tubulin | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Breast Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| 10mM PBS with 0.05% BSA | |
| 203068 | |
| IgG2b κ | |
| Protein A or G purified |
| TUBB/4470 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta | |
| Recombinant full-length human beta Tubulin protein (TUBB5) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title