missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
beta Tubulin Monoclonal antibody specifically detects beta Tubulin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | beta Tubulin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | TUBB/4470 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Formulation | 10mM PBS with 0.05% BSA |
| Gene Alias | beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?