missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-2 Adrenergic R/ADRB2 Antibody (4A6C9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61711-0.025ml
This item is not returnable.
View return policy
Description
beta-2 Adrenergic R/ADRB2 Monoclonal specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| beta-2 Adrenergic R/ADRB2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:100000 | |
| ADRBR, adrenergic, beta-2-, receptor, surface, B2ARADRB2R, BAR, beta-2 adrenergic receptor, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor | |
| Mouse | |
| 65.4 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 4A6C9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ADRB2 | |
| Purified recombinant fragment of human ADRB2 (AA: 302-413) expressed in E. Coli. | |
| Protein G purified | |
| Asthma, GPCR, Immunology, Neuroscience, Phospho Specific, Signal Transduction | |
| 154 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction