missing translation for 'onlineSavingsMsg'
Learn More

beta-2 Adrenergic R/ADRB2 Antibody (4A6C9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

217.00€ - 552.00€

Specifications

Antigen beta-2 Adrenergic R/ADRB2
Clone 4A6C9
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:100000
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18243534
View Documents
Novus Biologicals
NBP2-61711
0.1 mL
552.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18650689
View Documents
Novus Biologicals
NBP2-61711-0.025ml
0.025 mL
217.00€
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

beta-2 Adrenergic R/ADRB2 Monoclonal specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

beta-2 Adrenergic R/ADRB2
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Unconjugated
Mouse
ADRBR, adrenergic, beta-2-, receptor, surface, B2ARADRB2R, BAR, beta-2 adrenergic receptor, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor
ADRB2
IgG1
Protein G purified
4A6C9
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:100000
Monoclonal
Purified
Asthma, GPCR, Immunology, Neuroscience, Phospho Specific, Signal Transduction
154
Purified recombinant fragment of human ADRB2 (AA: 302-413) expressed in E. Coli.
Primary
65.4 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.