missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta 2 Adaptin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74371-0.025mg
This item is not returnable.
View return policy
Description
Beta 2 Adaptin Polyclonal specifically detects Beta 2 Adaptin in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot.
Specifications
| Beta 2 Adaptin | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 1 mg/ml BSA with 0.05% Sodium Azide | |
| AP2B1 | |
| Synthetic peptide corresponding to residues I(588) H R K H L P I H H G S T D A G D S P(606) of human beta Adaptin 2. | |
| 0.025 mg | |
| Primary | |
| beta-2 Adaptin | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:500 | |
| Adapter-related protein complex 2 beta subunit, Adaptor protein complex AP-2 subunit beta, adaptor-related protein complex 2, beta 1 subunit, ADTB2adaptin, beta 2 (beta), AP105B, AP2-BETA, Beta-2-adaptin, Beta-adaptin, CLAPB1AP-2 complex subunit beta, Clathrin assembly protein complex 2 beta large chain, clathrin-associated/assembly/adaptor protein, large, beta 1, DKFZp781K0743, Plasma membrane adaptor HA2/AP2 adaptin beta subunit | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 163 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction