missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta 2 Adaptin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
195.00€
Specifications
| Antigen | Beta 2 Adaptin |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18514130
|
Novus Biologicals
NB100-74371-0.025mg |
0.025 mg | |||||||
Description
Beta 2 Adaptin Polyclonal specifically detects Beta 2 Adaptin in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot.Specifications
| Beta 2 Adaptin | |
| Western Blot 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Adapter-related protein complex 2 beta subunit, Adaptor protein complex AP-2 subunit beta, adaptor-related protein complex 2, beta 1 subunit, ADTB2adaptin, beta 2 (beta), AP105B, AP2-BETA, Beta-2-adaptin, Beta-adaptin, CLAPB1AP-2 complex subunit beta, Clathrin assembly protein complex 2 beta large chain, clathrin-associated/assembly/adaptor protein, large, beta 1, DKFZp781K0743, Plasma membrane adaptor HA2/AP2 adaptin beta subunit | |
| AP2B1 | |
| IgG | |
| Affinity Purified | |
| beta-2 Adaptin |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS containing 1 mg/ml BSA with 0.05% Sodium Azide | |
| 163 | |
| Synthetic peptide corresponding to residues I(588) H R K H L P I H H G S T D A G D S P(606) of human beta Adaptin 2. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title