missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAT3/BAG6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38537-25ul
This item is not returnable.
View return policy
Description
BAT3/BAG6 Polyclonal specifically detects BAT3/BAG6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| BAT3/BAG6 | |
| Polyclonal | |
| Western Blot 1:100 - 1:250, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| P46379 | |
| BAG6 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: APETNAPNHPSPAEYVEVLQELQRLESRLQPFLQRYYEVLGAAATTDYNNNHEGREEDQRLINLVGESLRLLGNTFVALSDLRCNLACTPPRHLHVVRPMSHYTT | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| BAT3HLA-B associated transcript-3, BCL2-associated athanogene 6BAG-6, D6S52EHLA-B-associated transcript 3, G3BAG family molecular chaperone regulator 6, HLA-B associated transcript 3, large proline-rich protein BAT3, Protein G3, Protein Scythe, scythe | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 7917 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction