missing translation for 'onlineSavingsMsg'
Learn More

BAT3/BAG6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

369.00€ - 593.00€

Specifications

Antigen BAT3/BAG6
Dilution Western Blot 1:100 - 1:250, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18107498
View Documents
Novus Biologicals
NBP2-38537
0.1 mL
593.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18684376
View Documents
Novus Biologicals
NBP2-38537-25ul
25 μL
369.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BAT3/BAG6 Polyclonal specifically detects BAT3/BAG6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

BAT3/BAG6
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
BAT3HLA-B associated transcript-3, BCL2-associated athanogene 6BAG-6, D6S52EHLA-B-associated transcript 3, G3BAG family molecular chaperone regulator 6, HLA-B associated transcript 3, large proline-rich protein BAT3, Protein G3, Protein Scythe, scythe
BAG6
IgG
Affinity Purified
Western Blot 1:100 - 1:250, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
Polyclonal
Rabbit
Human
P46379
7917
This antibody was developed against a recombinant protein corresponding to amino acids: APETNAPNHPSPAEYVEVLQELQRLESRLQPFLQRYYEVLGAAATTDYNNNHEGREEDQRLINLVGESLRLLGNTFVALSDLRCNLACTPPRHLHVVRPMSHYTT
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.