missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAP1 Mouse anti-Human, Clone: BAP1/2431, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | BAP1 |
|---|---|
| Clone | BAP1/2431 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Protein Array |
| Applications | ELISA, Peptide Array |
Description
BAP1 Monoclonal specifically detects BAP1 in Human samples. It is validated for ELISA, Protein Array.Specifications
| BAP1 | |
| 0.2 mg/mL | |
| ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| 8314 | |
| IgG2c κ | |
| Protein A or G purified |
| BAP1/2431 | |
| ELISA, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2 | |
| Recombinant human BAP1 protein fragment (around aa 191-326) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title