missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAP1 Mouse anti-Human, Clone: BAP1/2431, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07390-100ug
This item is not returnable.
View return policy
Description
BAP1 Monoclonal specifically detects BAP1 in Human samples. It is validated for ELISA, Protein Array.
Specifications
| BAP1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Protein Array | |
| BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2 | |
| Recombinant human BAP1 protein fragment (around aa 191-326) (exact sequence is proprietary) | |
| 100 μg | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors | |
| 8314 | |
| Store at 4°C. | |
| IgG2c κ |
| ELISA, Peptide Array | |
| BAP1/2431 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion