missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATPB Antibody (4.3E8.D10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
200.00€
Specifications
| Antigen | ATPB |
|---|---|
| Clone | 4.3E8.D10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 - 2 μg/mL, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunoprecipitation 2 - 5 μg, In vitro assay |
| Applications | Western Blot |
Description
ATPB Monoclonal specifically detects ATPB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, In vitro assay.Specifications
| ATPB | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| ATP synthase subunit beta, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide, ATPMB, ATPSBmitochondrial ATP synthase beta subunit, EC 3.6.3, EC 3.6.3.14, MGC5231, mitochondrial ATP synthetase, beta subunit | |
| ATP5F1B | |
| IgG1 | |
| Immunogen affinity purified | |
| Detects the beta subunit of ATP synthase from mouse rat and human samples. This is useful as a mitochondrial marker. |
| 4.3E8.D10 | |
| Western Blot 1 - 2 μg/mL, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunoprecipitation 2 - 5 μg, In vitro assay | |
| Monoclonal | |
| Purified | |
| RUO | |
| P06576 | |
| 506 | |
| Intact rat mitochondria. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title