missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATPB Antibody (4.3E8.D10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB600-1171-0.025mg
This item is not returnable.
View return policy
Description
ATPB Monoclonal specifically detects ATPB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, In vitro assay.
Specifications
| ATPB | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 - 2 μg/mL, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunoprecipitation 2 - 5 μg, In vitro assay | |
| ATP synthase subunit beta, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide, ATPMB, ATPSBmitochondrial ATP synthase beta subunit, EC 3.6.3, EC 3.6.3.14, MGC5231, mitochondrial ATP synthetase, beta subunit | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 506 | |
| Human | |
| Purified |
| Western Blot | |
| 4.3E8.D10 | |
| Unconjugated | |
| P06576 | |
| ATP5F1B | |
| Intact rat mitochondria. | |
| 0.025 mg | |
| Primary | |
| Detects the beta subunit of ATP synthase from mouse rat and human samples. This is useful as a mitochondrial marker. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction