missing translation for 'onlineSavingsMsg'
Learn More

ATP6V1G1 Antibody - Azide and BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody

381.00€

Specifications

Antigen ATP6V1G1
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18550734
View Documents
Novus Biologicals
NB110-13819-0.025mg
0.025 mg
381.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ATP6V1G1 Polyclonal specifically detects ATP6V1G1 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot.
Specifications

Specifications

ATP6V1G1
Western Blot 1:100-1:2000
Polyclonal
Chicken
Human
ATP6G1V-ATPase 13 kDa subunit 1, ATP6GATP6J, ATP6GL, ATPase, H+ transporting, lysosomal (vacuolar proton pump), member J, ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G isoform 1, ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G1, DKFZp547P234, vacuolar ATP synthase subunit M16, vacuolar H(+)-ATPase subunit G 1, Vacuolar proton pump subunit G 1, Vacuolar proton pump subunit M16, V-ATPase subunit G 1, Vma10, V-type proton ATPase subunit G 1
ATP6V1G1
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
1 mg/mL
Western Blot
Unconjugated
RUO
PBS with No Preservative
9550
Synthetic peptide: corresponding to amino acids 1-118 of Human ATP6J.
Affinity Purified
ATP6J
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.