missing translation for 'onlineSavingsMsg'
Learn More

ATP6V1G1 Antibody - Azide and BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody

Brand:  Novus Biologicals NB110-13819-0.025mg

 View more versions of this product

Product Code. 18550734

  • 381.00€ / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ATP6V1G1 Polyclonal specifically detects ATP6V1G1 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATP6V1G1
Polyclonal
Unconjugated
PBS with No Preservative
ATP6V1G1
Synthetic peptide: corresponding to amino acids 1-118 of Human ATP6J.
0.025 mg
Primary
ATP6J
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Western Blot
1 mg/mL
Western Blot 1:100-1:2000
ATP6G1V-ATPase 13 kDa subunit 1, ATP6GATP6J, ATP6GL, ATPase, H+ transporting, lysosomal (vacuolar proton pump), member J, ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G isoform 1, ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G1, DKFZp547P234, vacuolar ATP synthase subunit M16, vacuolar H(+)-ATPase subunit G 1, Vacuolar proton pump subunit G 1, Vacuolar proton pump subunit M16, V-ATPase subunit G 1, Vma10, V-type proton ATPase subunit G 1
Chicken
Affinity Purified
RUO
9550
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only